missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC62 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81813AF700
This item is not returnable.
View return policy
Description
SEC62 Polyclonal antibody specifically detects SEC62 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SEC62 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| Dtrp1, hTP-1, S.cerevisiae, homolog of, SEC62 homolog (S. cerevisiae), TLOC1, TP-1, translocation protein 1, translocation protein SEC62 | |
| SEC62 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SEC62. The immunogen is located within amino acids 270 - 320 of SEC62. Amino Acid Squence: TYTHEYKGPKADLKKDEKS | |
| 0.1 mL | |
| Signal Transduction | |
| 7095 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction