missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC61B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93984-0.1ml
This item is not returnable.
View return policy
Description
SEC61B Polyclonal antibody specifically detects SEC61B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Gene Knock-Out
Specifications
| SEC61B | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Knockout Validated | |
| protein translocation complex beta, protein transport protein SEC61 beta subunit, protein transport protein Sec61 subunit beta, Sec61 beta subunit, Sec61 complex, beta subunit, SEC61B | |
| A synthetic peptide corresponding to a sequence within amino acids 1-96 of human SEC61B (NP_006799.1). MPGPTPSGTNVGSSGRSPSKAVAARAAGSTVRQRKNASCGTRSAGRTTSAGTGGMWRFYTEDSPGLKVGPVPVLVMSLLFIASVFMLHIWGKYTRS | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones | |
| 10952 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Gene Knock-Out | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction