missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SEC61alpha Recombinant Rabbit Monoclonal Antibody (K01_1Y19)

Product Code. 30829713
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829713 50 μL
30828448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829713 Supplier Invitrogen™ Supplier No. MA564899
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: SKTWIEVSGS SAKDVAKQLK EQQMVMRGHR DTSMVHELNR YIPTA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 98%, Pig - 100%, Cynomolgus monkey - 100%.

The SEC61 complex is a major component of the endoplasmic reticulum (ER) translocation site. This complex is composed of three subunits: SEC61alpha, SEC61beta and SEC61gamma. SEC61alpha is a polytopic integral membrane protein. It is a core component of the ER translocation complex, playing a crucial role in the insertion of secretory and membrane polypeptides into the ER.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC61alpha
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1Y19
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene SEC61A2
Gene Accession No. Q9H9S3, Q9JLR1
Gene Alias protein transport protein Sec61 subunit alpha; protein transport protein Sec61 subunit alpha isoform 2; Sec61 alpha; Sec61 alpha 2 subunit; Sec61 alpha 2 subunit (S. cerevisiae); Sec61 alpha form 2; sec61 alpha-2; Sec61 translocon alpha 2 subunit; Sec61, alpha subunit 2; Sec61, alpha subunit 2 (S. cerevisiae); SEC61A2
Gene Symbols SEC61A2
Host Species Rabbit
Immunogen Recombinant protein of human SEC61A (aa 376-420).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361273, 55176, 57743
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.