missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEC61alpha Recombinant Rabbit Monoclonal Antibody (9W7I3)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538005
This item is not returnable.
View return policy
Description
Positive Samples: 293T, SH-SY5Y, Mouse lung, Mouse testis, Mouse brain, Rat testis Immunogen sequence: SMGAIFEDPV HVVVYIIFML GSCAFFSKTW IEVSGSSAKD VAKQLKEQQM VMRGHRDTSM VHELNRYIPT AAAFGGLCIG ALSVLADFLG AIGSGTGILL A.
The SEC61 complex is a major component of the endoplasmic reticulum (ER) translocation site. This complex is composed of three subunits: SEC61alpha, SEC61beta and SEC61gamma. SEC61alpha is a polytopic integral membrane protein. It is a core component of the ER translocation complex, playing a crucial role in the insertion of secretory and membrane polypeptides into the ER.
Specifications
| SEC61alpha | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H9S3, Q9JLR1 | |
| SEC61A2 | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human Sec61A (Q9H9S3). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 9W7I3 | |
| Unconjugated | |
| SEC61A2 | |
| protein transport protein Sec61 subunit alpha; protein transport protein Sec61 subunit alpha isoform 2; Sec61 alpha; Sec61 alpha 2 subunit; Sec61 alpha 2 subunit (S. cerevisiae); Sec61 alpha form 2; sec61 alpha-2; Sec61 translocon alpha 2 subunit; Sec61, alpha subunit 2; Sec61, alpha subunit 2 (S. cerevisiae); SEC61A2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 361273, 55176, 57743 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction