missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC61A Rabbit anti-Human, Mouse, Rat, Clone: ARC2323, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16599-20UL
This item is not returnable.
View return policy
Description
SEC61A Monoclonal antibody specifically detects SEC61A in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SEC61A | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| ARC2323 | |
| Western Blot 1:500 - 1:2000 | |
| HSEC61, protein transport protein SEC61 alpha subunit, protein transport protein Sec61 subunit alpha, protein transport protein Sec61 subunit alpha isoform 1, SEC61, Sec61 alpha 1 subunit (S. cerevisiae), Sec61 alpha-1, sec61 homolog, SEC61A | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human SEC61A (Q9H9S3). Sequence: SMGAIFEDPVHVVVYIIFMLGSCAFFSKTWIEVSGSSAKDVAKQLKEQQMVMRGHRDTSMVHELNRYIPTAAAFGGLCIGALSVLADFLGAIGSGTGILLA | |
| 20 μg | |
| Membrane Trafficking and Chaperones | |
| 29927 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction