missing translation for 'onlineSavingsMsg'
Learn More

SEC61A Antibody, Novus Biologicals™

Product Code. 18364850 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364850 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364850 Supplier Novus Biologicals Supplier No. NBP286799

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SEC61A Polyclonal specifically detects SEC61A in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC61A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias HSEC61, protein transport protein SEC61 alpha subunit, protein transport protein Sec61 subunit alpha, protein transport protein Sec61 subunit alpha isoform 1, SEC61, Sec61 alpha 1 subunit (S. cerevisiae), Sec61 alpha-1, sec61 homolog, SEC61A
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human SEC61A
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 29927
Test Specificity This antibody made against SEC61A1 but the immunogen shows 84% sequence identity with human SEC61A2. Reactivity with SEC61A2 has not been determined experimentally.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.