missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC5/EXOC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12751-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and the functions of the exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. This interaction has been shown to mediate filopodia formation in fibroblasts.Specifications
| SEC5/EXOC2 | |
| Polyclonal | |
| Unconjugated | |
| EXOC2 | |
| EXOC2, exocyst complex component 2, Exocyst complex component Sec5, SEC5, SEC5L1, Sec5p | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171455, 55770, 66482 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O54921, Q96KP1, Q9D4H1 | |
| Exoc2 | |
| SEC5/EXOC2 Fusion Protein Ag3147 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction