missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEC31A Monoclonal Antibody (5D12C6)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531900
This item is not returnable.
View return policy
Description
MA5-31900 has been tested in indirect ELISA.
The protein encoded by this gene is similar to yeast Sec31 protein. Yeast Sec31 protein is known to be a component of the COPII protein complex which is responsible for vesicle budding from endoplasmic reticulum (ER). This protein was found to colocalize with SEC13, one of the other components of COPII, in the subcellular structures corresponding to the vesicle transport function. An immunodepletion experiment confirmed that this protein is required for ER-Golgi transport. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| SEC31A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O94979 | |
| SEC31A | |
| Purified recombinant fragment of human SEC31A (AA: 429-571) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 5D12C6 | |
| Unconjugated | |
| SEC31A | |
| 1810024J13Rik; ABP125; ABP130; AU042160; cb354; HSPC275; HSPC334; KIAA0905; LOW QUALITY PROTEIN: protein transport protein Sec31A; protein transport protein Sec31A; SEC31 homolog A (S. cerevisiae); SEC31 homolog A, COPII coat complex component; SEC31 homolog A, COPII coating complex component; sec31a; SEC31L1; SEC31-like 1; SEC31-like protein 1; sec31p; sec31p homolog; SEC31-related protein A; Unknown (protein for MGC:63547); VAP1; vesicle associated protein; vesicle-associated protein 1; Web1-like protein; yeast Sec31p homolog; zgc:63547 | |
| Mouse | |
| Protein G | |
| RUO | |
| 22872 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction