missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC31A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | SEC31A |
|---|---|
| Dilution | Western Blot 1:1000 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SEC31A Polyclonal antibody specifically detects SEC31A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SEC31A | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 22872 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ABP125ABP130KIAA0905Web1-like protein, DKFZp686N07171, HSPC334, MGC90305, protein transport protein Sec31A, protein-transport protein SEC31, SEC31 homolog A (S. cerevisiae), SEC31L1, SEC31-like 1, SEC31-like 1 (S. cerevisiae), SEC31-like protein 1, SEC31-related protein A, yeast Sec31p homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-500 of human SEC31A (NP_055748.2). PTNTQWCFDIQWCPRNPAVLSAASFDGRISVYSIMGGSTDGLRQKQVDKLSSSFGNLDPFGTGQPLPPLQIPQQTAQHSIVLPLKKPPKWIRRPVGASFSFGGKLVTFENVRMPSHQGAEQQQQQHHVFISQVVTEKEFLSRSDQLQQAVQSQGFINYCQKKIDASQTEFEKNVWSFLKVNFEDDSRGKYLELLGYRKEDL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title