missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEC24D Recombinant Rabbit Monoclonal Antibody (23GB2510), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | SEC24D |
|---|---|
| Clone | 23GB2510 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088963
![]() |
Invitrogen™
MA553568 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088264
|
Invitrogen™
MA553569 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SEC24D Recombinant Monoclonal antibody specifically detects SEC24D in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| SEC24D | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O94855 | |
| 310843, 69608, 9871 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2510 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2310020L09Rik; CLCRP2; Gm1349; KIAA0755; Protein transport protein Sec24D; SEC24 family member D; SEC24 family, member D; SEC24 homolog D, COPII coat complex component; SEC24 related gene family, member D; Sec24 related gene family, member D (S. cerevisiae); SEC24D; SEC24-related protein D | |
| A synthesized peptide derived from human SEC24D (1-43AA). | |
| Primary | |
| -20°C | |
| SEC24D |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title