missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SEC24D Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA565793

Product Code. 15978524

  • £473.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: IFLLANGLHMF LWLGVSSPPE LIQGIFNVPS FAHINTDMTL LPEVGNPYSQ QLRMIMGIIQ QKRPYSMKLT IVKQREQP Highest antigen sequence identity to the following orthologs - mouse 85%, rat 84%.

The protein encoded by this gene is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. This gene product is implicated in the shaping of the vesicle, and also in cargo selection and concentration.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SEC24D
Polyclonal
Unconjugated
SEC24D
2310020L09Rik; CLCRP2; Gm1349; KIAA0755; Protein transport protein Sec24D; SEC24 family member D; SEC24 family, member D; SEC24 homolog D, COPII coat complex component; SEC24 related gene family, member D; Sec24 related gene family, member D (S. cerevisiae); SEC24D; SEC24-related protein D
Rabbit
Antigen affinity chromatography
RUO
9871
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O94855
SEC24D
Recombinant Human SEC24D. Recombinant protein control fragment (Product #RP-106466).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.