Learn More
SEC24C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559101
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RLPSMTGPLL PGQSFGGPSV SQPNHVSSPP QALPPGTQMT GPLGPLPPMH SPQQPGYQPQ QNGSFGPARG PQSNYGGPYP AAPTF Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 78%.
Interacts with HSP70 and can stimulate its ATPase activity. Stimulates the association between HSC70 and HIP.Specifications
SEC24C | |
Polyclonal | |
Unconjugated | |
SEC24C | |
2610204K03Rik; KIAA0079; mKIAA0079; Protein transport protein Sec24C; SEC24 family member C; SEC24 family, member C; SEC24 homolog C, COPII coat complex component; SEC24 related gene family, member C; Sec24 related gene family, member C (S. cerevisiae); Sec24c; SEC24-related protein C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9632 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P53992 | |
SEC24C | |
Recombinant protein corresponding to Human SEC24C | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only