missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEC23IP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543797
This item is not returnable.
View return policy
Description
Peptide sequence: VPFIPVTQAS ASPASLLLPG EDSTDVGEED SFLGQTSIHT SAPQTFSYFS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.
SEC23IP is involved in protein transport from the endoplasmic reticulum to the Golgi apparatus. The protein encoded by this gene was identified by its interaction with a mouse protein similar to yeast Sec23p, an essential component of the COPII. This protein shares significant similarity with phospholipid-modifying proteins, especially phosphatidic acid preferring-phospholipase A1. Overexpression of this protein has been shown to cause disorganization of the endoplasmic reticulum-Golgi intermediate compartment and Golgi apparatus, which suggests its role in the early secretory pathway.
Specifications
| SEC23IP | |
| Polyclonal | |
| Unconjugated | |
| SEC23IP | |
| D7Ertd373e; MSTP053; P125; P125A; SEC23 interacting protein; SEC23-interacting protein; Sec23-interacting protein p125; SEC23-interacting protein-like protein; SEC23IP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11196 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y6Y8 | |
| SEC23IP | |
| Synthetic peptide directed towards the N-terminal of human SEC23IP (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction