missing translation for 'onlineSavingsMsg'
Learn More

SEC23IP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen SEC23IP
Dilution Western Blot 1:200 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30232263
View Documents
Novus Biologicals
NBP3-35544-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30232903
View Documents
Novus Biologicals
NBP3-35544-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SEC23IP Polyclonal antibody specifically detects SEC23IP in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications

Specifications

SEC23IP
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Unconjugated
Rabbit
Golgi Apparatus Markers
PBS (pH 7.3), 50% glycerol
11196
IgG
Affinity purified
Western Blot 1:200 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
Human, Mouse, Rat
P125, P125A, SEC23 interacting protein, SEC23-interacting protein
Recombinant fusion protein containing a sequence corresponding to amino acids 600-940 of human SEC23IP (NP_009121.1).,, Sequence:, NLSKCPGPLAVANGVVKQLHFQEKQMPEEPKLTLDESYDLVVENKEVLTLQETLEALSLSEYFSTFEKEKIDMESLLMCTVDDLKEMGIPLGPRKKIANFVEHKAAKLKKAASEKKAVAATSTKGQEQSAQKTKDMASLPSESNEPKRKLPVGACVSSVCVNYESFEVGAGQVSVAYNSLDFEPEIFFALGSPIAMFLTIRGVDRIDENYSLPTCKGFFNIYHPLDPVAYRLEPMIVPDLDLKAVLIPHHKGRKRLHLELKESLSRMGSDLKQGFISSLKSAWQTLNEFARAHTSSTQLQEELEKVANQIKEEEEKQVVEAEKVVESPDFSKDEDYLGKVG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.