missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sec23a Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519011
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, equine, human and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
The protein encoded by this gene is a member of the SEC23 subfamily of the SEC23/SEC24 family. It is part of a protein complex and found in the ribosome-free transitional face of the endoplasmic reticulum (ER) and associated vesicles. This protein has similarity to yeast Sec23p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The encoded protein is suggested to play a role in the ER-Golgi protein trafficking.
Specifications
| Sec23a | |
| Polyclonal | |
| Unconjugated | |
| Sec23a | |
| CLSD; hSec23A; MGC26267; Msec23; protein transport protein Sec23A; Sec23; Sec23 homolog A (S. cerevisiae); Sec23 homolog A, coat complex II component; Sec23 homolog A, COPII coat complex component; Sec23a; SEC23A (S. cerevisiae); Sec23r; SEC23-related protein A | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 20334 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q01405 | |
| Sec23a | |
| Synthetic peptide sequence (ANRAATTGHVID) corresponding to the internal amino acids of SEC23A (aa 335-347). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction