missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SEC23A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA588348

Product Code. 16516621

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MTTYLEFIQQ NEERDGVRFS WNVWPSSRLE ATRMVVPVAA LFTPLKERPD LPPIQYEPVL CSRTTCRAVL NPLCQVDYRA KLWACNFCYQ RNQFPPSYAG ISELNQPAEL LPQFSSIEYV VLRGPQMPLI FLYVVDTCME DEDLQALKES; Positive Samples: B-cell, LO2, HeLa, SGC-7901, HT-29, Mouse brain, Mouse liver, Rat brain; Cellular Location: Golgi apparatus membrane, Peripheral membrane protein, Smooth endoplasmic reticulum membrane.

The protein encoded by this gene is a member of the SEC23 subfamily of the SEC23/SEC24 family. It is part of a protein complex and found in the ribosome-free transitional face of the endoplasmic reticulum (ER) and associated vesicles. This protein has similarity to yeast Sec23p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The encoded protein is suggested to play a role in the ER-Golgi protein trafficking.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SEC23A
Polyclonal
Unconjugated
Sec23a
CLSD; hSec23A; MGC26267; Msec23; protein transport protein Sec23A; Sec23; Sec23 homolog A (S. cerevisiae); Sec23 homolog A, coat complex II component; Sec23 homolog A, COPII coat complex component; Sec23a; SEC23A (S. cerevisiae); Sec23r; SEC23-related protein A
Rabbit
Affinity Chromatography
RUO
10484, 20334, 58817
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.85 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q01405, Q15436
Sec23a
Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human Sec23A (NP_0063552).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.