missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC22B Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37701B
This item is not returnable.
View return policy
Description
SEC22B Polyclonal antibody specifically detects SEC22B in Human samples. It is validated for Western Blot
Specifications
| SEC22B | |
| Polyclonal | |
| Western Blot | |
| ERS24, ERS-24sec22b, SEC22 vesicle trafficking protein homolog B (S. cerevisiae), SEC22 vesicle trafficking protein homolog B (S. cerevisiae) (gene/pseudogene), SEC22 vesicle trafficking protein-like 1, SEC22 vesicle trafficking protein-like 1 (S. cerevisiae), SEC22 vesicle-trafficking protein homolog B, SEC22 vesicle-trafficking protein-like 1, SEC22, vesicle trafficking protein (S. cerevisiae)-like 1, SEC22L1ER-Golgi SNARE of 24 kDa, vesicle-trafficking protein SEC22b | |
| This antibody maps to a region between residue 100 to 150 of human SEC22 Vesicle Trafficking Protein Homolog B using the numbering given in entry NP_004883.3 (GeneID 9554). | |
| 0.1 mL | |
| Signal Transduction | |
| 9554 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction