missing translation for 'onlineSavingsMsg'
Learn More

SEC22B Antibody (1E1), Novus Biologicals™

Product Code. 18368248 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368248 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368248 Supplier Novus Biologicals Supplier No. H00009554M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SEC22B Monoclonal antibody specifically detects SEC22B in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC22B
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1E1
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004883
Gene Alias ERS24, ERS-24sec22b, SEC22 vesicle trafficking protein homolog B (S. cerevisiae), SEC22 vesicle trafficking protein homolog B (S. cerevisiae) (gene/pseudogene), SEC22 vesicle trafficking protein-like 1, SEC22 vesicle trafficking protein-like 1 (S. cerevisiae), SEC22 vesicle-trafficking protein homolog B, SEC22 vesicle-trafficking protein-like 1, SEC22, vesicle trafficking protein (S. cerevisiae)-like 1, SEC22L1ER-Golgi SNARE of 24 kDa, vesicle-trafficking protein SEC22b
Host Species Mouse
Immunogen SEC22L1 (NP_004883, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MVLLTMIARVADGLPLAASMQEDEQSGRDLQQYQSQAKQLFRKLNEQSPTRCTLEAGAMTFHYIIEQGVCYLVLCEAAFPKKLAFAYLEDLHSEFDEQHGKKVPTVSRPY
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9554
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.