missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SEC14L3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA563727

Product Code. 15711502

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VEVLLPDEGM QKYDKELTPV Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 90%.

The protein encoded by this gene is highly similar to the protein encoded by the Saccharomyces cerevisiae SEC14 gene. The SEC14 protein is a phophatidylinositol transfer protein that is essential for biogenesis of Golgi-derived transport vesicles, and thus is required for the export of yeast secretory proteins from the Golgi complex. The specific function of this protein has not yet been determined. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SEC14L3
Polyclonal
Unconjugated
SEC14L3
1110069O07Rik; 45 kDa secretory protein; rsec45; SEC14 like lipid binding 3; Sec14l3; SEC14-like 3; SEC14-like 3 (S. cerevisiae); SEC14-like lipid binding 3; SEC14-like protein 3; SEC14p-like protein TAP2; secretory protein, 45kD; Spf2; TAP2; Tocopherol-associated protein 2
Rabbit
Antigen affinity chromatography
RUO
266629
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9UDX4
SEC14L3
Recombinant protein corresponding to Human SEC14L3. Recombinant protein control fragment (Product #RP-101921).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.