Learn More
Invitrogen™ SEC13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521339
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (87%), Xenopus laevis (88%), Chicken (94%), Rhesus Monkey (98%), Chimpanzee (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene belongs to the SEC13 family of WD-repeat proteins. It is a constituent of the endoplasmic reticulum and the nuclear pore complex. It has similarity to the yeast SEC13 protein, which is required for vesicle biogenesis from endoplasmic reticulum during the transport of proteins. Multiple alternatively spliced transcript variants have been found.
Specifications
| SEC13 | |
| Polyclonal | |
| Unconjugated | |
| SEC13 | |
| 1110003H02Rik; D3S1231E; GATOR complex protein SEC13; npp-20; Protein SEC13 homolog; Sec13; SEC13 homolog; SEC13 homolog (S. cerevisiae); SEC13 homolog, nuclear pore and COPII coat complex component; SEC13 homolog, nuclear pore and COPII coating complex component; SEC13 related; SEC13A; Sec13l1; SEC13-like 1; SEC13-like 1 isoform; SEC13-like protein 1; SEC13R; SEC13-related protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6396 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P55735 | |
| SEC13 | |
| Recombinant protein encompassing a sequence within the center region of human SEC13L1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.