missing translation for 'onlineSavingsMsg'
Learn More

sec-Butylbenzene, TRC

Product Code. 30338824
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338824 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338824 Supplier Toronto Research Chemicals Supplier No. TRCB6927051G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material sec-Butylbenzene
CAS 135-98-8
Molecular Formula C10 H14
Quantity 1 g
Synonym Benzene, sec-butyl- (8CI), (1-Methylpropyl)benzene, (RS)-2-Phenylbutane, (±)-sec-Butylbenzene, (α-Methylpropyl)benzene, 2-Phenylbutane, NSC 8466, sec-Butylbenzene
InChI Formula InChI=1S/C10H14/c1-3-9(2)10-7-5-4-6-8-10/h4-9H,3H2,1-2H3
SMILES CCC(C)c1ccccc1
IUPAC Name butan-2-ylbenzene
Molecular Weight (g/mol) 134.22
Formula Weight 134.1096
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.