missing translation for 'onlineSavingsMsg'
Learn More

sec-Butylbenzene, 99%

Product Code. 11473980
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473980 25 mL 25mL
11436116 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473980 Supplier Thermo Scientific Alfa Aesar Supplier No. B22962.AC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 135-98-8 | C10H14 | 134.222 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-98-8
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00009329
InChI Key ZJMWRROPUADPEA-UHFFFAOYSA-N
Synonym sec-butylbenzene, 2-phenylbutane, 1-methylpropyl benzene, secondary butylbenzene, benzene, 1-methylpropyl, s-butylbenzene, benzene, sec-butyl, 1-sec-butylbenzene, methylpropyl benzene, secbutylbenzene
PubChem CID 8680
ChEBI CHEBI:35097
IUPAC Name butan-2-ylbenzene
SMILES CCC(C)C1=CC=CC=C1

Specifications

Melting Point -75°C
Density 0.862
Boiling Point 173°C to 174°C
Flash Point 52°C (125°F)
Odor Aromatic
Refractive Index 1.49
Quantity 25 mL
UN Number UN2709
Beilstein 1903902
Merck Index 14,1550
Formula Weight 134.22
Percent Purity 99%
Chemical Name or Material sec-Butylbenzene
Show More Show Less
compliance-icons
Product Identifier
  • sec-Butylbenzene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: sec-Butylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.