missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sec-Butylamine, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 2-Butylamine |
| CAS | 13952-84-6 |
| Molecular Formula | C4 H11 N |
| Quantity | 25 g |
| Synonym | sec-Butylamine (6CI,8CI), (RS)-sec-butylamine, (±)-2-Aminobutane, (±)-2-Butanamine, (±)-2-Butylamine, (±)-sec-Butylamine, 1-Methylpropanamine, 1-Methylpropylamine, 2-AB, 2-Aminobutane, 2-Butylamine, Butafume, Butan-2-ylamine, DL-sec-Butylamine, NSC 8030, Tutane, dl-2-Butylamine |
| InChI Formula | InChI=1S/C4H11N/c1-3-4(2)5/h4H,3,5H2,1-2H3 |
| SMILES | CCC(C)N |
| IUPAC Name | butan-2-amine |
| Molecular Weight (g/mol) | 73.14 |
| Formula Weight | 73.0891 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction