missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sec-Butyl Methyl Ether, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 2-Methoxybutane |
| CAS | 6795-87-5 |
| Molecular Formula | C5 H12 O |
| Quantity | 500 mg |
| Synonym | Butane, 2-methoxy-, Ether, sec-butyl methyl (6CI,7CI,8CI), 2-Methoxybutane, (±)-2-Methoxybutane, (±)-Methyl 2-butyl ether, 2-Butyl methyl ether, Methyl 1-methylpropyl ether, Methyl sec-butyl ether, sec-Butyl methyl ether, sec-Butyl methyl ether |
| InChI Formula | InChI=1S/C5H12O/c1-4-5(2)6-3/h5H,4H2,1-3H3 |
| SMILES | CCC(C)OC |
| IUPAC Name | 2-methoxybutane |
| Molecular Weight (g/mol) | 88.15 |
| Formula Weight | 88.0888 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction