missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl methyl ether, 99%

Product Code. 15450228
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450228 5 mL 5mL
15460228 25 mL 25mL
15470228 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15450228 Supplier Thermo Scientific Alfa Aesar Supplier No. H53427.AA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6795-87-5 | C5H12O | 88.15 g/mol

sec-Butyl methyl ether is used as a solvent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
sec-Butyl methyl ether is used as a solvent in organic synthesis.

Solubility
Miscible with water, alcohol, ether and acetone.

Notes
Incompatible with strong oxidizing agents. Avoid conditions like flames, sparks and direct sunlight.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6795-87-5
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00190205
InChI Key FVNIMHIOIXPIQT-UHFFFAOYNA-N
Synonym sec-butyl methyl ether, butane, 2-methoxy, methyl sec-butyl ether, ether, sec-butyl methyl, 2-butyl methyl ether, methyl 1-methylpropyl ether, s-butyl methyl ether, butane, 2-methoxy-9ci, butane,2-methoxy, acmc-1b64a
PubChem CID 23238
IUPAC Name 2-methoxybutane
SMILES CCC(C)OC

Specifications

Density 0.742
Boiling Point 57°C
Flash Point −30°C (−22°F)
Refractive Index 1.372
Quantity 5 mL
UN Number UN2350
Solubility Information Miscible with water,alcohol,ether and acetone.
Formula Weight 88.15
Percent Purity 99%
Chemical Name or Material sec-Butyl methyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.