missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl Mercaptan, TRC

Product Code. 30338948
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338948 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338948 Supplier Toronto Research Chemicals Supplier No. TRCB6926905G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material sec-Butyl Mercaptan
CAS 513-53-1
Molecular Formula C4H10S
Quantity 5 g
Synonym (±)-2-Butanethiol, 1-Methyl-1-propanethiol, 2-Butyl mercaptan, 2-Butylthiol, 2-Mercaptobutane, 3-Methyl-2-propanethiol, NSC 78417, s-Butyl mercaptan, sec-Butanethiol, sec-Butyl mercaptan, sec-Butyl thioalcohol, sec-Butylthiol, 2-Butanethiol, (±)-2-Butanethiol, 1-Methyl-1-propanethiol, 2-Butyl mercaptan, 2-Butylthiol, 2-Mercaptobutane, 3-Methyl-2-propanethiol, NSC 78417, s-Butyl mercaptan, sec-Butanethiol, sec-Butyl thioalcohol, sec-Butylthiol, 2-Butanethiol
InChI Formula InChI=1S/C4H10S/c1-3-4(2)5/h4-5H,3H2,1-2H3
SMILES CCC(C)S
IUPAC Name butane-2-thiol
Molecular Weight (g/mol) 90.19
Formula Weight 90.0503
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.