missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10077632
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4426-79-3
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.19
MDL Number MFCD00041108
InChI Key TUFJIDJGIQOYFY-UHFFFAOYNA-N
Synonym sec-butyl isothiocyanate, 2-butyl isothiocyanate, butane, 2-isothiocyanato, isothiocyanic acid sec-butyl ester, isothiocyanic acid, sec-butyl ester, butan-2-isothiocyanate, 2-butylisothiocyanate, s-butyl isothiocyanate, 2-isothiocyanato-butane
PubChem CID 78151
IUPAC Name 2-isothiocyanatobutane
SMILES CCC(C)N=C=S

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 115.2
Chemical Name or Material sec-Butyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'