missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sec-Butyl Acetate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Acetic acid-sec.-butyl ester |
| CAS | 105-46-4 |
| Molecular Formula | C6 H12 O2 |
| Quantity | 1 g |
| Synonym | Acetic acid, sec-butyl ester (8CI), sec-Butyl alcohol acetate (6CI), sec-Butyl alcohol, ester with AcOH (4CI), (±)-2-Butyl acetate, (±)-sec-Butyl acetate, 1-Methylpropyl acetate, 2-Acetoxybutane, 2-Butyl acetate, NSC 8034, Ultrasolve M 1200, sec-Butyl acetate, sec-Butyl ethanoate |
| InChI Formula | InChI=1S/C6H12O2/c1-4-5(2)8-6(3)7/h5H,4H2,1-3H3 |
| SMILES | CCC(C)OC(=O)C |
| IUPAC Name | butan-2-yl acetate |
| Molecular Weight (g/mol) | 116.16 |
| Formula Weight | 116.0837 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction