missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl acetate, 98%

Product Code. p-7046305
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11383297

Brand: Thermo Scientific Alfa Aesar L13892.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105-46-4 | C6H12O2 | 116.16 g/mol

sec-Butyl acetate is used in organic synthesis, catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
sec-Butyl acetate is used in organic synthesis, catalytic agent, petrochemical additive.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-46-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009328
InChI Key DCKVNWZUADLDEH-UHFFFAOYNA-N
Synonym sec-butyl acetate, dl-sec-butyl acetate, 2-butyl acetate, acetic acid, 1-methylpropyl ester, 1-methylpropyl acetate, sec-butyl ethanoate, 1-methylpropyl ethanoate, acetic acid, sec-butyl ester, 2-butanol acetate, sec-butyl alcohol acetate
PubChem CID 7758
IUPAC Name butan-2-yl acetate
SMILES CCC(C)OC(C)=O

Specifications

Melting Point -99°C
Density 0.872
Boiling Point 111°C to 112°C
Flash Point 16°C (60°F)
Refractive Index 1.389
Quantity 25 g
UN Number UN1123
Beilstein 1720689
Merck Index 14,1536
Solubility Information Slightly soluble in water.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material sec-Butyl acetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.