missing translation for 'onlineSavingsMsg'
Learn More

Sebaconitrile, 98%

Product Code. 10197940 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10197940 25 g Glass bottle 25g
10567752 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10197940 Supplier Thermo Scientific Acros Supplier No. 419250250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1871-96-1 | C10H16N2 | 164.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1871-96-1
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.25
MDL Number MFCD00019900
InChI Key DFJYZCUIKPGCSG-UHFFFAOYSA-N
Synonym sebaconitrile, 1,8-dicyanooctane, octamethylene dicyanide, oktamethylendikyanid, oktamethylendikyanid czech, octane-1,8-dicarbonitrile, sebacic acid dinitrile, acmc-209epg, wln: nc8cn, 4-02-00-02089 beilstein handbook reference
PubChem CID 74639
IUPAC Name decanedinitrile
SMILES C(CCCCC#N)CCCC#N

Specifications

Melting Point 7.0°C
Color Colorless to Yellow
Density 0.9100g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula NC(CH2)8CN
Refractive Index 1.4470 to 1.4490
Quantity 25 g
Specific Gravity 0.91
Formula Weight 164.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Sebaconitrile, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Sebaconitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.