missing translation for 'onlineSavingsMsg'
Learn More

Sebaconitrile, 95%

Product Code. 11416186
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416186 5 g 5g
11426186 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416186 Supplier Thermo Scientific Alfa Aesar Supplier No. B23010.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1871-96-1 | C10H16N2 | 164.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1871-96-1
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD00019900
InChI Key DFJYZCUIKPGCSG-UHFFFAOYSA-N
Synonym sebaconitrile, 1,8-dicyanooctane, octamethylene dicyanide, oktamethylendikyanid, oktamethylendikyanid czech, octane-1,8-dicarbonitrile, sebacic acid dinitrile, acmc-209epg, wln: nc8cn, 4-02-00-02089 beilstein handbook reference
PubChem CID 74639
IUPAC Name decanedinitrile
SMILES C(CCCCC#N)CCCC#N

Specifications

Melting Point 4°C to 8°C
Density 0.91
Boiling Point 199°C to 200°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1750739
Formula Weight 164.25
Percent Purity 95%
Chemical Name or Material Sebaconitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.