missing translation for 'onlineSavingsMsg'
Learn More

Sebacic Acid, TRC

Product Code. 30426029
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 30426029

Brand: Toronto Research Chemicals TRCS21141010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,8-Octanedicarboxylic acid (Decanedioic acid)
CAS 111-20-6
Purity Grade Notes HPLC
Molecular Formula C10 H18 O4
Quantity 10 g
Synonym Decanedioic acid (9CI','ACI), Sebacic acid (8CI), 1,10-Decanedioic acid, 1,8-Octanedicarboxylic acid, n-Decanedioic acid, NSC 19492, NSC 97405
InChI Formula InChI=1S/C10H18O4/c11-9(12)7-5-3-1-2-4-6-8-10(13)14/h1-8H2,(H,11,12)(H,13,14)
SMILES OC(=O)CCCCCCCCC(=O)O
IUPAC Name decanedioic acid
Molecular Weight (g/mol) 202.25
Formula Weight 202.1205
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.