missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sebacic Acid, TRC
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Specifications
Specifications
| Chemical Name or Material | 1,8-Octanedicarboxylic acid (Decanedioic acid) |
| CAS | 111-20-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 H18 O4 |
| Quantity | 1 g |
| Synonym | Decanedioic acid (9CI','ACI), Sebacic acid (8CI), 1,10-Decanedioic acid, 1,8-Octanedicarboxylic acid, n-Decanedioic acid, NSC 19492, NSC 97405 |
| InChI Formula | InChI=1S/C10H18O4/c11-9(12)7-5-3-1-2-4-6-8-10(13)14/h1-8H2,(H,11,12)(H,13,14) |
| SMILES | OC(=O)CCCCCCCCC(=O)O |
| IUPAC Name | decanedioic acid |
| Molecular Weight (g/mol) | 202.25 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction