missing translation for 'onlineSavingsMsg'
Learn More

Sebacic Acid, 98%

Product Code. 10073853 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
10kg
1kg
2.5kg
This item is not returnable. View return policy

Product Code. 10073853

Brand: Thermo Scientific Acros 132630100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-20-6 | C10H18O4 | 202.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-20-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00004440
InChI Key CXMXRPHRNRROMY-UHFFFAOYSA-N
Synonym sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
PubChem CID 5192
ChEBI CHEBI:41865
IUPAC Name decanedioic acid
SMILES C(CCCCC(=O)O)CCCC(=O)O

Specifications

Melting Point 131.0°C to 134.0°C
Color White
Boiling Point 294.4°C (100.0 mmHg)
Flash Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic drum
Linear Formula HO2C(CH2)8CO2H
Quantity 10 kg
Beilstein 02, IV, 2078
Merck Index 15, 8552
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: 0.1% in ether (17°C), freely soluble in alcohol, esters and ketones, sparingly soluble in hydrocarbons, insoluble in ethyl ether and carbon tetrachloride
Formula Weight 202.25
Percent Purity 98%
Physical Form Powder or Granules
Chemical Name or Material Sebacic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.