missing translation for 'onlineSavingsMsg'
Learn More

Sebacic acid, 98+%

Product Code. 11412207
Click to view available options
Quantity:
100 g
250 g
1000 g
5000 g
Unit Size:
1000g
100g
250g
5000g
This item is not returnable. View return policy

Product Code. 11412207

Brand: Thermo Scientific Alfa Aesar A14158.0I

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-20-6 | C10H18O4 | 202.25 g/mol

Sebacic acid is used as a monomer for nylon 610, plasticizers, lubricants, hydraulic fluids, cosmetics and candles. It is also used as intermediates in antiseptics, aromatics and painting materials. It is used in the synthesis of poly(glycerol sebacate) and novel bio-nylon. It finds application as adhesives, corrosion inhibitors and solvents. Its derivative, diethyl Sebacate is used in plastics manufacturing and engineering, as well as drug delivery procedures through the use of biodegradable encapsulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sebacic acid is used as a monomer for nylon 610, plasticizers, lubricants, hydraulic fluids, cosmetics and candles. It is also used as intermediates in antiseptics, aromatics and painting materials. It is used in the synthesis of poly(glycerol sebacate) and novel bio-nylon. It finds application as adhesives, corrosion inhibitors and solvents. Its derivative, diethyl Sebacate is used in plastics manufacturing and engineering, as well as drug delivery procedures through the use of biodegradable encapsulations.

Solubility
Slightly soluble in water.

Notes
Incompatible with bases, reducing agents and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-20-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00004440
InChI Key CXMXRPHRNRROMY-UHFFFAOYSA-N
Synonym sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
PubChem CID 5192
ChEBI CHEBI:41865
IUPAC Name decanedioic acid
SMILES C(CCCCC(=O)O)CCCC(=O)O

Specifications

Melting Point 131°C to 134°C
Density 1.271
Boiling Point 295°C (100 mmHg)
Flash Point 220°C (428°F)
Linear Formula HO2C(CH2)8CO2H
Quantity 5000 g
Beilstein 1210591
Merck Index 14,8415
Solubility Information Slightly soluble in water.
Formula Weight 202.25
Percent Purity ≥98%
Chemical Name or Material Sebacic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.