missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Seasonal H1N1 Nucleocapsid Protein Polyclonal antibody specifically detects Seasonal H1N1 Nucleocapsid Protein in Influenza Virus N1, Virus samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Seasonal H1N1 Nucleocapsid Protein |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Nucleocapsid protein, Nucleoprotein, Protein N |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a synthetic peptide from the seasonal H1N1 NP protein. The immunogen is located within amino acids 350 - 400 of Seasonal H1N1 Nucleocapsid Protein. Amino Acid Squence: NMDNMGSS |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?