missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Clear identification via numbers in sequence
- Integrated control label with the same number, for accompanying documents
- Tear-off perforation for each label
- Once closed with seal-it, then the container cannot be opened without destroying the seal-it label
- White labelling field on seal-it can be written on with all conventional writing tools
Specifications
Specifications
| Color | Red |
| Product Type | Security seal |
| For Use With (Application) | Retention Samples |
| Includes | Numbers in sequence, integrated control label, tear-off perforation for each label |
| Quantity | 500/Roll |
| Dimensions (L x W) | 178 x 30 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?