missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Seal Crimper
Secure tear-away aluminum seals.
£1070.00
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
15193065
|
Labconco™
7578000 |
13 mm |
£1070.00
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
15103075
|
Labconco™
7578100 |
20 mm |
£1070.00
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
Description
- Secure tear-away Aluminum Seals on Serum Bottles
- Available in 13mm or 20mm corkage