missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SEA 0400
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SEA 0400 is a potent Na+/Ca2+ exchanger (NCX) inhibitor. Inhibits NCX activity in rat astrocytes, microglia and cultured neurons, canine cardiac sarcolemmal vesicles and rat cardiomyocytes (IC50 values are 5 and 8.3, 33, 90 and 92 nM, respectively). Reduces infarct volumes and exhibits protective effects against myocardial ischemia in rats.
Specifications
Specifications
| Product Type | Inhibitor |
| Target Proteins | Na+/Ca2+ Exchanger |
| CAS | 223104-29-8 |
| Molecular Formula | C21H19F2NO3 |
| Formula Weight | 371.38g/mol |
| Content And Storage | Store at -20°C |
| For Use With (Application) | Na+/Ca2+ Exchanger Inhibitors |
| Quantity | 10 mg |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction