missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Se-NADA

Product Code. 18758566
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18758566 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18758566 Supplier Tocris Bioscience™ Supplier No. 7449/5
  • ONLINE EXCLUSIVE PRICE
    £328.00
    £309.75 / 5mg
    Save £18.25
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Orange fluorescent benzoselenadiazole D-amino acid (FDAA) for imaging bacteria; also photosensitizer

Se-NADA is an orange fluorescent benzoselenadiazole D-amino acid (FDAA) for imaging bacteria. It is covalently incorporated into peptidoglycans in the cell walls of live bacteria. It also acts as a photosensitizer, bringing about bacterial cell death on exposure to visible light. Se-NADA is effective against both Gram-positive and Gram-negative bacteria. Ex/Em = 470/590 nm.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-((7-Nitro-2,1,3-benzoselenadiazol-4-yl)amino)-D-alanine hydrochloride
CAS 2571574-70-2
Quantity 5 mg
Target Fluorescent Probes
Molecular Formula C9H9N5O4Se.HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.