missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Se-methylseleno-L-cysteine, 98%

Product Code. 10096383 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10096383 50 mg Glass Bottle 50mg
10125370 250 mg Glass Bottle 250mg
10167463 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10096383 Supplier Thermo Scientific Chemicals Supplier No. 300640500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26046-90-2 | C4H9NO2Se | 182.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26046-90-2
Molecular Formula C4H9NO2Se
Molecular Weight (g/mol) 182.092
MDL Number MFCD00800565
InChI Key XDSSPSLGNGIIHP-VKHMYHEASA-N
Synonym methylselenocysteine, se-methylselenocysteine, se-methyl-l-selenocysteine, 3-methylseleno-l-alanine, selenium-methylselenocystine, r-2-amino-3-methylselanyl propanoic acid, se-methyl-seleno-l-cysteine, unii-twk220499z, ccris 5465, l-alanine, 3-methylseleno
PubChem CID 147004
ChEBI CHEBI:27812
IUPAC Name (2R)-2-amino-3-methylselanylpropanoic acid
SMILES C[Se]CC(C(=O)O)N

Specifications

Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 50 mg
Formula Weight 182.08
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Se-methylseleno-L-cysteine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.