missing translation for 'onlineSavingsMsg'
Learn More

SDZ 220-581, MedChemExpress

Product Code. p-200099068
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16495518

Brand: Medchem Express HY13059

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SDZ 220-581 is an orally active, potent, competitive NMDA receptor antagonist with pKi value of 7.7.

SDZ 220-581 is an orally active, potent, competitive NMDA receptor antagonist with pKi value of 7.7.

Specifications

Chemical Name or Material SDZ 220-581
Physical Form Solid
CAS 174575-17-8
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H17ClNO5P
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information DMSO : 8.57 mg/mL (23.18 mM; Need ultrasonic and warming)
SMILES ClC1=CC=CC=C1C2=CC(C[C@H](N)C(O)=O)=CC(CP(O)(O)=O)=C2
Molecular Weight (g/mol) 369.74
Formula Weight 369.74
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.