missing translation for 'onlineSavingsMsg'
Learn More

SDZ 220-581 hydrochloride, MedChemExpress

Product Code. 16451697
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451697 50 mg 50mg
16415528 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451697 Supplier Medchem Express Supplier No. HY13059B
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SDZ 220-581 hydrochloride is an orally active, potent, competitive NMDA receptor antagonist with pKi value of 7.7.

SDZ 220-581 hydrochloride is an orally active, potent, competitive NMDA receptor antagonist with pKi value of 7.7.

Specifications

Chemical Name or Material SDZ 220-581 hydrochloride
Physical Form Solid
CAS 179411-93-9
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H18Cl2NO5P
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information DMSO : 5 mg/mL (12.31 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES ClC1=CC=CC=C1C2=CC(C[C@H](N)C(O)=O)=CC(CP(O)(O)=O)=C2.[H]Cl
Molecular Weight (g/mol) 406.2
Formula Weight 406.2
Percent Purity 99.6%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.