missing translation for 'onlineSavingsMsg'
Learn More

SDZ 220-581 Ammonium salt, MedChemExpress

Product Code. 16441697
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441697 50 mg 50mg
16405528 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16441697 Supplier Medchem Express Supplier No. HY13059A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress SDZ 220-581 Ammonium salt is an orally active, potent, competitive NMDA receptor antagonist with pKi value of 7.7.

SDZ 220-581 Ammonium salt is an orally active, potent, competitive NMDA receptor antagonist with pKi value of 7.7.

Specifications

Chemical Name or Material SDZ 220-581 Ammonium salt
Physical Form Solid
CAS 179411-94-0
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H20ClN2O5P
For Use With (Application) Neuroscience-Neurodegeneration
SMILES ClC1=CC=CC=C1C2=CC(C[C@H](N)C(O)=O)=CC(CP(O)(O)=O)=C2.N
Molecular Weight (g/mol) 386.77
Formula Weight 386.77
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.