missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDZ 220-581 Ammonium salt, MedChemExpress
Description
SDZ 220-581 Ammonium salt is an orally active, potent, competitive NMDA receptor antagonist with pKi value of 7.7.
Specifications
Specifications
| Chemical Name or Material | SDZ 220-581 Ammonium salt |
| Physical Form | Solid |
| CAS | 179411-94-0 |
| Color | Off-White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C16H20ClN2O5P |
| For Use With (Application) | Neuroscience-Neurodegeneration |
| SMILES | ClC1=CC=CC=C1C2=CC(C[C@H](N)C(O)=O)=CC(CP(O)(O)=O)=C2.N |
| Molecular Weight (g/mol) | 386.77 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?