missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDSL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35574-20ul
This item is not returnable.
View return policy
Description
SDSL Polyclonal antibody specifically detects SDSL in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| SDSL | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| EC 4.3.1.17, EC 4.3.1.19, L-serine deaminase, L-serine dehydratase/L-threonine deaminase, L-threonine dehydratase, SDH, SDH 2, SDS-RS1, Serine dehydratase 2, serine dehydratase related sequence 1, serine dehydratase-like, TDH | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-137 of human SDSL (NP_612441.1).,, Sequence:, MDGPVAEHAKQEPFHVVTPLLESWALSQVAGMPVFLKCENVQPSGSFKIRGIGHFCQEMAKKGCRHLVCSSGGNAGIAAAYAARKLGIPATIVLPESTSLQVVQRLQGEGAEVQLTGKVWDEANLRAQELAKRDGWE | |
| 20 μL | |
| metabolism | |
| 113675 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction