missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDSL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35574-100ul
This item is not returnable.
View return policy
Description
SDSL Polyclonal antibody specifically detects SDSL in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| SDSL | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| EC 4.3.1.17, EC 4.3.1.19, L-serine deaminase, L-serine dehydratase/L-threonine deaminase, L-threonine dehydratase, SDH, SDH 2, SDS-RS1, Serine dehydratase 2, serine dehydratase related sequence 1, serine dehydratase-like, TDH | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-137 of human SDSL (NP_612441.1).,, Sequence:, MDGPVAEHAKQEPFHVVTPLLESWALSQVAGMPVFLKCENVQPSGSFKIRGIGHFCQEMAKKGCRHLVCSSGGNAGIAAAYAARKLGIPATIVLPESTSLQVVQRLQGEGAEVQLTGKVWDEANLRAQELAKRDGWE | |
| 100 μL | |
| metabolism | |
| 113675 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion