missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SDS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570549
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
SDS3 is a subunit of the histone deacetylase.
Specifications
| SDS3 | |
| Polyclonal | |
| Unconjugated | |
| SUDS3 | |
| 2400003N08Rik; 2410008L21Rik; 45 kDa Sin3-associated polypeptide; AU067672; FLJ00052; mSds3; RGD1305986; SAP45; SDS3; SDS3 homolog, SIN3A corepressor complex component; sin3 histone deacetylase corepressor complex component SDS3; Sin3 histone deacetylase corepressor complex component SDS3-like protein; sin3A-associated protein, 45kDa; SUDS3; suppressor of defective silencing 3 homolog; suppressor of defective silencing 3 homolog (S. cerevisiae); suppressor of defective silencing 3 protein homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 71954 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8BR65 | |
| SUDS3 | |
| synthetic peptide directed towards the N-terminal region of mouse SDS3. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction