missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SDS3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA559807

Product Code. 15797651

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: KAAVKEFEDK KVELKENLIA ELEEKKKMIE NEKLTMELTG DSMEVKPIMT RKLRRRPNDP VPIPDKRRKP APAQLNYLLT DEQIMEDLRT LNKLKSPKRP A Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

SDS3 is a subunit of the histone deacetylase.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SDS3
Polyclonal
Unconjugated
SUDS3
2400003N08Rik; 2410008L21Rik; 45 kDa Sin3-associated polypeptide; AU067672; FLJ00052; mSds3; RGD1305986; SAP45; SDS3; SDS3 homolog, SIN3A corepressor complex component; sin3 histone deacetylase corepressor complex component SDS3; Sin3 histone deacetylase corepressor complex component SDS3-like protein; sin3A-associated protein, 45kDa; SUDS3; suppressor of defective silencing 3 homolog; suppressor of defective silencing 3 homolog (S. cerevisiae); suppressor of defective silencing 3 protein homolog
Rabbit
Antigen affinity chromatography
RUO
64426
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9H7L9
SUDS3
Recombinant protein corresponding to Human SDS3. Recombinant protein control fragment (Product #RP-97715).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.