missing translation for 'onlineSavingsMsg'
Learn More

Melford SDS, Powder

Product Code. 30018375
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
25 kg
Unit Size:
100g
1kg
250g
25kg
500g
5kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018375 250 g 250g
30018374 1 kg 1kg
30018376 25 kg 25kg
30018373 100 g 100g
30018378 5 kg 5kg
30018377 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30018375 Supplier Melford Supplier No. L22010250.0
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ionic detergent for denaturing proteins prior to gel electrophoresis.

Ionic detergent for denaturing proteins prior to gel electrophoresis.

Specifications

Chemical Name or Material SDS, Powder
CAS 151-21-3
Quantity 250 g
Molecular Formula C12H25NaO4S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.